missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENOPH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532096
This item is not returnable.
View return policy
Description
Recommended positive controls: THP-1, HL-60. Predicted reactivity: Mouse (93%), Rat (92%), Pig (91%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Bifunctional enzyme that catalyzes the enolization of 2,3-diketo-5-methylthiopentyl-1-phosphate (DK-MTP-1-P) into the intermediate 2-hydroxy-3-keto-5-methylthiopentenyl-1-phosphate (HK-MTPenyl-1-P), which is then dephosphorylated to form the acireductone 1,2-dihydroxy-3-keto-5-methylthiopentene (DHK-MTPene).
Specifications
| ENOPH1 | |
| Polyclonal | |
| Unconjugated | |
| ENOPH1 | |
| 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; 2,3-diketo-5-methylthio-1-phosphopentane phosphatase {ECO:0000255; 2310057D15Rik; acireductone synthase; BB183658; C81437; E1; E-1 enzyme; enolase-phosphatase 1; enolase-phosphatase E1; enolase-phosphatase E1 {ECO:0000255; ENOPH1; HAMAP-Rule:MF_03117}; Masa; MASA homolog; MASA homolog {ECO:0000255; MST145; MSTP145; mtnC; RGD1309016 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58478 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UHY7 | |
| ENOPH1 | |
| Recombinant protein fragment corresponding to a region within amino acids 1 and 244 of Human ENOPH1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction