Learn More
ENPEP Monoclonal Antibody (OTI1D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA503914S
Encompass_Preferred
 View more versions of this product
Product Code. 17500312
  • € 242.00 / 30µL
Description
ENPEP Monoclonal antibody specifically detects ENPEP in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
ENPEP
Monoclonal
0.94 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15848
Mouse
Affinity chromatography
RUO
9370
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Flow Cytometry, Western Blot
OTI1D1
Unconjugated
ADIPOQ
6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51
Human recombinant protein fragment corresponding to amino acids 19-244 of human ADIPOQ produced in HEK293T cell.