missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP-7/Alk-SMase Antibody (1), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89694PE
This item is not returnable.
View return policy
Description
ENPP-7/Alk-SMase Monoclonal antibody specifically detects ENPP-7/Alk-SMase in Human samples. It is validated for ELISA
Specifications
| ENPP-7/Alk-SMase | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA | |
| Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, Intestinal alkaline sphingomyelinase, MGC50179, NPP-7 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ENPP-7/Alk-SMase (Accession#: NP_848638.2; Met 1-Ser 439). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 339221 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction