missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP-7/Alk-SMase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99660-50ul
This item is not returnable.
View return policy
Description
ENPP-7/Alk-SMase Polyclonal antibody specifically detects ENPP-7/Alk-SMase in Human samples. It is validated for Western Blot, ELISA
Specifications
| ENPP-7/Alk-SMase | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:5000-1:10000 | |
| Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, Intestinal alkaline sphingomyelinase, MGC50179, NPP-7 | |
| Produced in rabbits immunized with purified, recombinant Human ENPP-7/Alk-SMase (Accession#: NP_848638.2; Met 1-Ser 439) | |
| 50 μg | |
| Lipid and Metabolism | |
| 339221 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction