missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENPP5 Recombinant Rabbit Monoclonal Antibody (JE62-55)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544817
This item is not returnable.
View return policy
Description
ENPP5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.
Specifications
| ENPP5 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9EQG7, Q9UJA9 | |
| ENPP5 | |
| Recombinant protein within human ENPP5 aa 378-477/477. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| JE62-55 | |
| Unconjugated | |
| ENPP5 | |
| AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 59084, 83965 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction