missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ENPP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532024
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: MCF-7. Predicted reactivity: Mouse (94%), Rat (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Choline-specific glycerophosphodiester phosphodiesterase. Hydrolyzes the classical substrate for phospholipase C, p-nitrophenyl phosphorylcholine, while it does not hydrolyze the classical nucleotide phosphodiesterase substrate, p-nitrophenyl thymidine 5'-monophosphate. Hydrolyzes lysophosphatidylcholine (LPC) to form monoacylglycerol and phosphorylcholine but not lysophosphatidic acid, showing it has a lysophospholipase C activity. Has a preference for LPC with short (12:0 and 14:0) or polyunsaturated (18:2 and 20:4) fatty acids. Also hydrolyzes glycerophosphorylcholine and sphingosylphosphorylcholine efficiently.
Specifikationer
| ENPP6 | |
| Polyclonal | |
| Unconjugated | |
| Enpp6 | |
| 4833421B01Rik; B830047L21Rik; choline-specific glycerophosphodiester phosphodiesterase; D8Ertd514e; ectonucleotide pyrophosphatase/phosphodiesterase 6; ectonucleotide pyrophosphatase/phosphodiesterase family member 6; E-NPP 6; Enpp6; E-NPP6; Glycerophosphocholine cholinephosphodiesterase; GPC-Cpde; Npp6; NPP-6; UNQ1889/PRO4334 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 133121 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6UWR7 | |
| Enpp6 | |
| Recombinant protein encompassing a sequence within the center region of human ENPP6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering