Learn More
ENT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32223
This item is not returnable.
View return policy
Description
ENT1 Polyclonal specifically detects ENT1 in Human samples. It is validated for Immunoprecipitation.
Specifications
| ENT1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| ENT1es-type, equilibrative nucleoside transporter 1, MGC1465, MGC3778, solute carrier family 29 (nucleoside transporters), member 1, Solute carrier family 29 member 1, solute carrier family 29, member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2030 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2 - 10 ug/mg lysate | |
| NP_001071645.1 | |
| SLC29A1 | |
| The immunogen this antibody was made to, maps to a region between residue 343 to 393 of human Equilibrative NBMPR-Sensitive Nucleoside Transporter using the numbering given in entry NP_001071645.1 (GeneID 2030). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |