Learn More
ENT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32223
Product Code. 18180165
  • € 583.00 / 100µL
Description
ENT1 Polyclonal specifically detects ENT1 in Human samples. It is validated for Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ENT1
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
ENT1es-type, equilibrative nucleoside transporter 1, MGC1465, MGC3778, solute carrier family 29 (nucleoside transporters), member 1, Solute carrier family 29 member 1, solute carrier family 29, member 1
Rabbit
Affinity Purified
RUO
2030
Store at 4C. Do not freeze.
Immunoprecipitation
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg lysate
NP_001071645.1
SLC29A1
The immunogen this antibody was made to, maps to a region between residue 343 to 393 of human Equilibrative NBMPR-Sensitive Nucleoside Transporter using the numbering given in entry NP_001071645.1 (GeneID 2030).