missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENT2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-81984F
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ENT2 Polyclonal antibody specifically detects ENT2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
| ENT2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| Delayed-early response protein 12, DER12Solute carrier family 29 member 2, ENT2, Equilibrative NBMPR-insensitive nucleoside transporter, Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleosidetransporter, equilibrative nucleoside transporter 2, HNP3636 kDa nucleolar protein HNP36, Hydrophobic nucleolar protein, 36 kDa, hydrophobic nucleolar protein, 36kD, Nucleoside transporter, ei-type, solute carrier family 29 (nucleoside transporters), member 2 | |
| ENT2 antibody was raised against a 19 amino acid peptide near the center of human ENT2. The immunogen is located within amino acids 250 - 300 of ENT2. Amino Acid Squence: KEPESEPDEPQKPGKPSV | |
| 0.1 mL | |
| Cancer | |
| 3177 | |
| Store at 4°C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido