missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterobacter aerogenes Antibody (57/82), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08869APC
This item is not returnable.
View return policy
Description
Enterobacter aerogenes Monoclonal antibody specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Enterobacter aerogenes | |
| Monoclonal | |
| APC | |
| Enterobacter aerogenes, Klebsiella mobilis | |
| A crude sonicate of Enterobacter aerogenes | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunofluorescence | |
| 57/82 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction