missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterobacter Cloacae Antibody (61/212), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62544B
This item is not returnable.
View return policy
Description
Enterobacter Cloacae Monoclonal specifically detects Enterobacter Cloacae in Bacteria samples. It is validated for ELISA.
Specifications
| Enterobacter Cloacae | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| This product recognizes Enterobacter cloacae, a gram-negative bacillus sometimes associated with urinary and respiratory tract infections in humans. | |
| Store at 4C in the dark. | |
| IgG2a |
| ELISA | |
| 61/212 | |
| ELISA | |
| Bacillus cloacae, Bacterium cloacae, Cloaca cloacae | |
| Enterobacter cloacae NCTC 10005 | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction