missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enteropeptidase/Enterokinase Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95274C
This item is not returnable.
View return policy
Description
Enteropeptidase/Enterokinase Polyclonal specifically detects Enteropeptidase/Enterokinase in Bovine samples. It is validated for ELISA.
Specifications
| Enteropeptidase/Enterokinase | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| ELISA | |
| DyLight 650 | |
| EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
| Produced in rabbits immunized with purified, recombinant Enteropeptidase/Enterokinase (NP_776864.1; Ile801-His1035). | |
| 0.1 mL | |
| Epitope Tags, Immunology, Innate Immunity | |
| 5651 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction