missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enteropeptidase/Enterokinase Antibody (MM0259- 8S12), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12411AF350
This item is not returnable.
View return policy
Description
Enteropeptidase/Enterokinase Monoclonal specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot.
Specifications
| Enteropeptidase/Enterokinase | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TMPRSS15 | |
| Human recombinant Enteropeptidase/Enterokinase | |
| Primary | |
| Detects human Enteropeptidase/Enterokinase heavy chain | |
| Store at 4°C in the dark. |
| Western Blot | |
| MM0259- 8S12 | |
| Western Blot | |
| EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
| Mouse | |
| 0.1 mL | |
| 5651 | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction