missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterovirus 71 3D Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33373-100ul
This item is not returnable.
View return policy
Description
Enterovirus 71 3D Monoclonal antibody specifically detects Enterovirus 71 3D in Human samples. It is validated for ELISA,Western Blot
Specifications
| Enterovirus 71 3D | |
| Monoclonal | |
| Western Blot 1:2000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL | |
| 3D, 3D polymerase, 3Dpol, EV71 3D, Protein 3D, RdRp, RNA-directed RNA polymerase | |
| Recombinant protein of Enterovirus 71 3D. | |
| 100 μL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction