missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Enterovirus 71 VP1 Recombinant Rabbit Monoclonal Antibody (HL1928)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547223
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Entervirus 71 is a potentially fatal virus first isolated in 1969. Enterovirus 71 has been found in the spinal cord and medulla and can produce a range of symptoms from rashes to meningitis and ecephalitis.
Specifications
| Enterovirus 71 VP1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| HL1928 | |
| Unconjugated | |
| Enterovirus; EV71 | |
| Recombinant fragment of Enterovirus 71 VP1. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction