Learn More
ENTPD1 Monoclonal Antibody (OTI2A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804587
Encompass_Preferred
 View more versions of this product
Product Code. 17574203
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.
Specifications
Specifications
ENTPD1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P49961
ENTPD1
Human recombit protein fragment corresponding to amino acids 38-307 of human ENTPD1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Western Blot
OTI2A1
Unconjugated
ENTPD1
ATPDase, CD39, NTPDase-1, SPG64
Mouse
Affinity Chromatography
RUO
953
-20° C, Avoid Freeze/Thaw Cycles