Learn More
ENTPD1 Mouse anti-Human, Clone: OTI2B10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804559
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.Specifications
| ENTPD1 | |
| Monoclonal | |
| Unconjugated | |
| ENTPD1 | |
| ATPDase, CD39, NTPDase-1, SPG64 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 953 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P49961 | |
| ENTPD1 | |
| Human recombit protein fragment corresponding to amino acids 38-307 of human ENTPD1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.