missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD1 Monoclonal Antibody (OTI3C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804742
This item is not returnable.
View return policy
Description
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.Specifications
| ENTPD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49961 | |
| ENTPD1 | |
| Human recombit protein fragment corresponding to amino acids 38-307 of human ENTPD1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3C2 | |
| Unconjugated | |
| ENTPD1 | |
| ATPDase, CD39, NTPDase-1, SPG64 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 953 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction