Learn More
Invitrogen™ ENTPD7 Polyclonal Antibody
GREENER_CHOICE
Product Code. 15586012
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15586012 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15586012 Supplier Invitrogen™ Supplier No. PA543366
Rabbit Polyclonal Antibody
Peptide sequence: EHRLKYQCFK SAWMYQVLHE GFHFPYDYPN LRTAQLVYDR EVQWTLGAIL Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 90%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
ENTPD7 is a multi-pass membrane protein. It belongs to the GDA1/CD39 NTPase family. It preferentially hydrolyzes nucleoside 5'-triphosphates. The order of activity with respect to possible substrates is UTP > GTP > CTP.
TRUSTED_SUSTAINABILITY
Specifications
Antigen ENTPD7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ENTPD7
Gene Accession No. Q9NQZ7
Gene Alias 1810012B13Rik; 1810020C02Rik; 2810003F23Rik; ectonucleoside triphosphate diphosphohydrolase 7; ENTPD7; Kiaa4066; Lalp1; Lysal2; lysosomal apyrase-like 2; lysosomal apyrase-like protein 1; NTPDase 7; RP11-483F11.1
Gene Symbols ENTPD7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ENTPD7 (aa 470-519).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57089
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG