missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ENTPD8 Polyclonal Antibody

Product Code. 16530542
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16530542 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16530542 Supplier Invitrogen™ Supplier No. PA597711

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ENTPD8 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

ENTPD8 is canalicular ectonucleoside NTPDase responsible for the main hepatic NTPDase activity. It is ectonucleoside NTPDases catalyze the hydrolysis of gamma-and beta-phosphate residues of nucleotides, playing a central role in concentration of extracellular nucleotides. ENTPD8 has activity toward ATP, ADP, UTP and UDP, but not toward AMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Entpd8
Gene Accession No. Q5MY95, Q8K0L2
Gene Alias ecto-ATP-diphosphohydrolase; ectonucleoside triphosphate diphosphohydrolase 8; ENTPD8; E-NTPDase; E-NTPDase 8; GLSR2492; I79_018892; liver ecto-ATP-diphosphohydrolase; NTPDase 8; NTPDase8; NTPDase-8; UNQ2492; UNQ2492/PRO5779
Gene Symbols Entpd8
Host Species Rabbit
Immunogen Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 8 protein (30-434AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377841, 72090
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.