Learn More
Memmert™ Entry Port 100 mm for IF750plus
Product Code. 17587225 Shop All Memmert Products
Change view
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17587225 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17587225 Supplier Memmert™ Supplier No. IF750plusF9
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 100 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IF750plus Incubator
Type Entry Port
Diameter (Metric) 100 mm