Learn More
Memmert™ Entry Port 23 mm for IF450plus, Custom TBC Right Side
Product Code. 17527045 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF450plus Incubator
Unit Size:
Each
Product Code. 17527045
Brand: Memmert™ IF450plusF5
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in right side
Specifications
For Use With (Equipment) IF450plus Incubator
Type Entry Port
Diameter (Metric) 23 mm