Learn More
Memmert™ Entry Port 23 mm for UF75plus, Standard Position Right Side Center/Center
Shop All Memmert Products
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position right side center/center
Specifications
Specifications
| For Use With (Equipment) | UF75plus Oven |
| Type | Entry Port |
| Diameter (Metric) | 23 mm |