Learn More
Memmert™ Entry Port 38 mm for IF30
Product Code. 17556915 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF30 Incubator
Unit Size:
Each
Product Code. 17556915
Brand: Memmert™ IF30F7
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IF30 Incubator
Type Entry Port
Diameter (Metric) 38 mm