Learn More
Memmert™ Entry Port 38 mm for IN450plus
Product Code. 17587775 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN450plus Incubator
Unit Size:
Each
Product Code. 17587775
Brand: Memmert™ IN450plusF7
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IN450plus Incubator
Type Entry Port
Diameter (Metric) 38 mm