Learn More
Memmert™ Entry Port 38 mm for IN450plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN450plus Incubator
Unit Size:
Each
Description
- Entry port with 38 mm clear diameter
- Flap and stopper closure
- Custom position TBC in the back
Specifications
Specifications
| For Use With (Equipment) | IN450plus Incubator |
| Type | Entry Port |
| Diameter (Metric) | 38 mm |