Learn More
Memmert™ Entry Port 38 mm for UF450
Product Code. 17549355 Shop All Memmert Products
Click to view available options
Diameter (Metric):
38 mm
Unit Size:
Each
Product Code. 17549355
Brand: Memmert™ UF450F7
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UF450 Oven
Type Entry Port
Diameter (Metric) 38 mm