missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation Envirogel GPC Cleanup Guard Column

Product Code. p-7236840
Encompass
Click to view available options
Solvents:
Cyclohexane/Ethyl Acetate
Methylene Chloride
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13107669

Brand: Waters Corporation 186001914

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Envirogel High-Resolution GPC cleanup columns are specifically designed to remove low volatility, high molecular weight interferences.

  • In the past, the cleanup procedure for environmental samples was performed on low-efficiency GPC columns based on packing particle diameters of 37-75 micron (200-400 mesh) Biobeads S-X resins.
  • The high-efficiency Envirogel Cleanup columns increase the speed of this process while simultaneously reducing solvent consumption.
  • They come packed in your choice of Methylene Chloride or a mixture of Cyclohexane/Ethyl acetate to minimize the need for risky solvent switchover.

Specifications

Quantity 1/Ea.
Particle Size 15 μm
Length (Metric) 30 mm
Type Guard Column
Diameter (Metric) 4.6 mm
Solvents Cyclohexane/Ethyl Acetate
Particle Shape Spherical
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.