missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EOGT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106975
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
The exact function of C3orf64 remains unknown.
Specifications
| EOGT | |
| Polyclonal | |
| Unconjugated | |
| Eogt | |
| A130022J15Rik; Aer61; AER61 glycosyltransferase; AI447490; AOS4; AW214473; AW259391; C3orf64; EGF domain specific O-linked N-acetylglucosamine transferase; EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase; EGF domain-specific O-linked N-acetylglucosamine transferase; EGF-O-GlcNAc transferase; EOGT; EOGT1; Extracellular O-linked N-acetylglucosamine transferase; glycosyltransferase Aer61 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 285203 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q5NDL2 | |
| Eogt | |
| Synthetic peptide directed towards the C-terminal region of human C3orf64. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction