missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EOMES Antibody (644730) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6166P
This item is not returnable.
View return policy
Description
EOMES Monoclonal antibody specifically detects EOMES in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| EOMES | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 644730 | |
| Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
| E. coli-derived recombinant human EOMES, Gly471-Pro686, Accession # O95936 | |
| 0.1 mL | |
| Innate Immunity | |
| 8320 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction