Learn More
PeproTech Eotaxin 2 Monoclonal Antibody, PeproTech®
Mouse Monoclonal Antibody
Brand: PeproTech 500-M31-500UG
Description
AA Sequence of recombinant protein: VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA. Preparation: Produced in BALB/c mice immunized with highly pure Recombinant Human Eotaxin-2 (CCL24). Anti-Human Eotaxin-2 (CCL24)-specific antibody was purified from ascites fluid by Protein A affinity chromatography.
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
Specifications
| Eotaxin 2 | |
| Monoclonal | |
| Unconjugated | |
| CCL24 | |
| CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6369 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| O00175 | |
| CCL24 | |
| E.coli-derived Recombinant Human Eotaxin-2 (CCL24). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.