missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epac2 Antibody (SR1071), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22082-50ul
This item is not returnable.
View return policy
Description
Epac2 Monoclonal antibody specifically detects Epac2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Epac2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1071 | |
| Western Blot 1:500-1:2000 | |
| CAMP-GEFII, cAMP-regulated guanine nucleotide exchange factor II, CGEF2EPAC 2, EPAC2, Exchange factor directly activated by cAMP 2, Exchange protein directly activated by cAMP 2, Nbla00496, putative protein product of Nbla00496, Rap guanine nucleotide exchange factor (GEF) 4, rap guanine nucleotide exchange factor 4, RAP guanine-nucleotide-exchange factor (GEF) 4 | |
| A synthesized peptide derived from human Epac2 (Uniprot #: Q8WZA2) | |
| 50 μg | |
| Signal Transduction | |
| 11069 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction