missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epac2 Antibody (SR1071), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22082-100ul
This item is not returnable.
View return policy
Description
Epac2 Monoclonal antibody specifically detects Epac2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Epac2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1071 | |
| Western Blot 1:500-1:2000 | |
| CAMP-GEFII, cAMP-regulated guanine nucleotide exchange factor II, CGEF2EPAC 2, EPAC2, Exchange factor directly activated by cAMP 2, Exchange protein directly activated by cAMP 2, Nbla00496, putative protein product of Nbla00496, Rap guanine nucleotide exchange factor (GEF) 4, rap guanine nucleotide exchange factor 4, RAP guanine-nucleotide-exchange factor (GEF) 4 | |
| A synthesized peptide derived from human Epac2 (Uniprot #: Q8WZA2) | |
| 100 μg | |
| Signal Transduction | |
| 11069 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?