Learn More
EPB42 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27443-1-AP-150UL
 View more versions of this product
Product Code. 18009465
  • € 410.00 / 150µL
Description
The EPB42 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EPB42. This antibody recognizes human antigen. The EPB42 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EPB42
350 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
E42P, EPB42, Erythrocyte protein 4.2, P4.2, PA, SPH5
150 μL
2038
Store at -20°C. Stable for one year after shipment.