Learn More
EPB42 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27443-1-AP-150UL
This item is not returnable.
View return policy
Description
The EPB42 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EPB42. This antibody recognizes human antigen. The EPB42 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| EPB42 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| E42P, EPB42, Erythrocyte protein 4.2, P4.2, PA, SPH5 | |
| 150 μL | |
| 2038 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |