Learn More
EPB42 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27443-1-AP-20UL
 View more versions of this product
Product Code. 18065015
  • € 145.00 / 20µL
Description
The EPB42 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EPB42. This antibody recognizes human antigen. The EPB42 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EPB42
350 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
E42P, EPB42, Erythrocyte protein 4.2, P4.2, PA, SPH5
20 μL
2038
Store at -20°C. Stable for one year after shipment.