missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCAM, anti-Human, Clone: EGP40/1556R, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00342.100ug
This item is not returnable.
View return policy
Specifications
| EPCAM | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS. | |
| 17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2 | |
| Original antibody is raised against recombinant protein corresponding to full length human EPCAM. | |
| RUO | |
| 4072 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| EGP40/1556R | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| epithelial cell adhesion molecule | |
| EPCAM | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction