Learn More
EPCAM Mouse anti-Human, CoraLite 594, Clone: 2A2D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66316-100UL
Description
The EPCAM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPCAM. This antibody recognizes human antigen. The EPCAM antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| EPCAM | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 4072 | |
| IgG2a |
| Immunofluorescence | |
| 2A2D5 | |
| Mouse | |
| Primary | |
| Human |