Learn More
EpCAM/TROP1 Antibody (G8.8R) [DyLight 350], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB8998D
Product Code. 18557677
  • € 678.00 / 0.10mL
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EpCAM/TROP1
Monoclonal
DyLight 350
50mM Sodium Borate
Rat
Protein A or G purified from cell culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, CyTOF
G8.8R
Flow Cytometry, CyTOF-reported
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
TE-71 Thymic epithelial cell line
0.1 mL
Cancer, Cancer Stem Cells, Tumor Biomarkers
4072
Store at 4°C in the dark.