missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM/TROP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 278.00 - € 456.00
Specifications
| Antigen | EpCAM/TROP1 |
|---|---|
| Concentration | 0.1mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18424261
|
Novus Biologicals
NBP1-89404-25ul |
25 μL |
€ 278.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18298007
|
Novus Biologicals
NBP1-89404 |
0.1 mL |
€ 456.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EpCAM/TROP1 Polyclonal specifically detects EpCAM/TROP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EpCAM/TROP1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P16422 | |
| 4072 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LFKAKQCNGTSMCWCVNTAGVRRTDKDTEITCSERVRTYWIIIELKHKAREKPYDSKSLRTALQKEITTRYQLDPKFITSILYENNVITIDLVQNSSQKTQNDVDIADVAYY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.1mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cancer Stem Cells, Tumor Biomarkers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| EPCAM | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title