missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCR Antibody (432714), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2749X
This item is not returnable.
View return policy
Description
EPCR Monoclonal antibody specifically detects EPCR in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| EPCR | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 432714 | |
| Flow Cytometry, CyTOF-ready | |
| APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial | |
| Mouse myeloma cell line NS0-derived recombinant mouse EPCR, Ala17-Ser214, Accession # Q64695 | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 10544 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction