missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPDR1 Antibody (11), Biotin , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06190B
This item is not returnable.
View return policy
Description
EPDR1 Monoclonal antibody specifically detects EPDR1 in Human samples. It is validated for ELISA
Specifications
| EPDR1 | |
| Monoclonal | |
| Biotin | |
| EPDR, ependymin related protein 1 (zebrafish), MERP1mammalian ependymin related protein 1, MERP-1Upregulated in colorectal cancer gene 1 protein, UCC1mammalian ependymin-related protein 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EPDR1 (Q9UM22-1; Met1-Trp224). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 54749 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction