missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPGN Antibody (RM0228-2L47), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12284AF594
This item is not returnable.
View return policy
Description
EPGN Monoclonal specifically detects EPGN in Mouse samples. It is validated for Western Blot.
Specifications
| EPGN | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EPGN | |
| Mouse recombinant EPGN | |
| 0.1 mL | |
| 255324 | |
| Store at 4°C in the dark. |
| Western Blot | |
| RM0228-2L47 | |
| Western Blot | |
| epigen, epithelial mitogen homolog (mouse), PRO9904, UNQ3072/PRO9904 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction