missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA2 Antibody (233720), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB639D
This item is not returnable.
View return policy
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| EphA2 | |
| Monoclonal | |
| DyLight 350 | |
| ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA2, Ala22-Ala535 (predicted), Accession # AAA82113 | |
| 0.5 mL | |
| Cancer, Protein Kinase | |
| 1969 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 233720 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction