missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA2 Antibody (301), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90650FR
This item is not returnable.
View return policy
Description
EphA2 Monoclonal specifically detects EphA2 in Mouse samples. It is validated for Flow Cytometry, ELISA.
Specifications
| EphA2 | |
| Monoclonal | |
| DyLight 680 | |
| ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 (NP_034269.2; Met1-Asn535). | |
| 0.1 mL | |
| Cancer, Protein Kinase | |
| 1969 | |
| Mouse | |
| Purified |
| Flow Cytometry, ELISA | |
| 301 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse EPHA4 Mouse EphA6 Mouse EPHA7 Human MEPHB1 Mouse EPHB3 Mouse EPHB4 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction