missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA2 Antibody (371805), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3035F
This item is not returnable.
View return policy
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-Out
Specifications
| EphA2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-Out | |
| 371805 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated | |
| ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
| Mouse myeloma cell line NS0-derived recombinant human EphA2, Gln25-Asn534, Accession # P29317 | |
| 0.1 mL | |
| Cancer, Protein Kinase | |
| 1969 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering