Learn More
Novus Biologicals™ EphA2 Antibody (Sanofi Aventis patent anti-EphA2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28354-50ug
 View more versions of this product
Product Code. 30044602
  • € 213.00 / 50µg
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EphA2
Monoclonal
Unconjugated
P29317
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Sanofi Aventis patent anti-EphA2
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
EphA2
50 μg
Cancer, Protein Kinase
1969