missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16347
This item is not returnable.
View return policy
Description
EphA4 Polyclonal specifically detects EphA4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| EphA4 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, TYRO1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1 | |
| Rabbit | |
| 110 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 2043 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000 | |
| P54764 | |
| EPHA4 | |
| Recombinant protein encompassing a sequence within the center region of human EphA4. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (99%), Chicken (93%), Pig (99%), Rat (99%), Xenopus tropicalis (85%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction