Learn More
EPHA5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55215-1-AP-20UL
 View more versions of this product
Product Code. 16886705
  • € 145.00 / 20µL
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
Specifications
Specifications
EPHA5
Polyclonal
Unconjugated
EPHA5
CEK7, EHK 1, EHK1, EK7, EPH homology kinase 1, EPH like kinase 7, EPH receptor A5, EPHA5, Ephrin type A receptor 5, HEK7, TYRO4
Rabbit
Antigen Affinity Chromatography
RUO
13839, 2044
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P54756, Q60629
EPHA5
Peptide
20 μL
Primary
Mouse, Human