Learn More
EphA6 Antibody (033), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90658MFV610
This item is not returnable.
View return policy
Description
EphA6 Monoclonal antibody specifically detects EphA6 in Mouse samples. It is validated for ELISA
Specifications
| EphA6 | |
| 033 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA6 (Accession#: NP_031964.2; Met 1-Gln 546). | |
| IgG |
| Monoclonal | |
| Rabbit | |
| Store at 4C in the dark. |