missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA6 Antibody (9J12), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-93605PCP
This item is not returnable.
View return policy
Description
EphA6 Monoclonal antibody specifically detects EphA6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EphA6 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 9J12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066 | |
| Mouse EphA6 (extracellular domain) | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 285220 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction