missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA6 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98326AF750
This item is not returnable.
View return policy
Description
EphA6 Polyclonal specifically detects EphA6 in Mouse samples. It is validated for ELISA.
Specifications
| EphA6 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Alexa Fluor 750 | |
| DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066 | |
| Produced in rabbits immunized with purified, recombinant EphA6 (NP_031964.2; Met 1-Gln 546). | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 285220 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction