missing translation for 'onlineSavingsMsg'
Learn More

EphA6 Monoclonal Antibody (1426CT591.205.91.119)
GREENER_CHOICE

Product Code. 17197248 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17197248 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17197248 Supplier Thermo Scientific Supplier No. MA537665

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EphA6 Monoclonal Antibody (1426CT591.205.91.119)

EphA6 Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA6
Applications Western Blot
Classification Monoclonal
Clone 1426CT591.205.91.119
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Epha6
Gene Accession No. Q9UF33
Gene Alias DKFZp434C1418; EHK2; EHK-2; EK12; EPA6; EPH homology kinase 2; Eph receptor A6; EPHA6; EPH-like kinase 12; ephrin receptor EphA6; ephrin type-A receptor 6; FLJ35246; HEK12; m-ehk2; PRO57066; tyrosine-protein kinase receptor EHK-2
Gene Symbols Epha6
Host Species Mouse
Immunogen EphA6 recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.